missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chondrolectin Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2576
This item is not returnable.
View return policy
Description
Chondrolectin Polyclonal specifically detects Chondrolectin in Human samples. It is validated for Western Blot.
Specifications
| Chondrolectin | |
| Polyclonal | |
| Unconjugated | |
| Q9H9P2 | |
| CHODL | |
| Mouse myeloma cell line NS0-derived recombinant human Chondrolectin Arg22-Asn216 Accession # Q9H9P2 | |
| 100 μg | |
| Primary | |
| Detects human Chondrolectin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| C21orf68, CHODL, chondrolectin, chromosome 21 open reading frame 68, FLJ12627, MT75, PRED12, Transmembrane protein MT75 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 140578 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction