missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chitinase 3-like 2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
£168.00 - £417.00
Specifications
| Antigen | Chitinase 3-like 2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18786357
![]() |
R&D Systems
AF5116-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18040222
|
R&D Systems
AF5116 |
100 μg |
£417.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chitinase 3-like 2 Polyclonal specifically detects Chitinase 3-like 2 in Human samples. It is validated for Western Blot.Specifications
| Chitinase 3-like 2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| CHI3L2, Chitinase 3 like 2, chitinase 3-like 2, chitinase-3-like protein 2, Chondrocyte protein 39, YKL39, YKL-39YKL39 | |
| CHI3L2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q15782 | |
| 1117 | |
| Mouse myeloma cell line NS0-derived recombinant human Chitinase 3-like 2 Tyr27-Leu390 Accession # Q15782 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Chitinase 3-like 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human CHIT-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title