missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chitinase 3-like 2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5116-SP
This item is not returnable.
View return policy
Description
Chitinase 3-like 2 Polyclonal specifically detects Chitinase 3-like 2 in Human samples. It is validated for Western Blot.
Specifications
| Chitinase 3-like 2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHI3L2, Chitinase 3 like 2, chitinase 3-like 2, chitinase-3-like protein 2, Chondrocyte protein 39, YKL39, YKL-39YKL39 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1117 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q15782 | |
| CHI3L2 | |
| Mouse myeloma cell line NS0-derived recombinant human Chitinase 3-like 2 Tyr27-Leu390 Accession # Q15782 | |
| 25 μg | |
| Primary | |
| Detects human Chitinase 3-like 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human CHIT-1 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction