Learn More
Human Chitinase 3-like 2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand:  R&D Systems AF5116
 View more versions of this product
Product Code. 18040222
  • £417.00 / 100µg
Description
Chitinase 3-like 2 Polyclonal specifically detects Chitinase 3-like 2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chitinase 3-like 2
Polyclonal
Unconjugated
Q15782
CHI3L2
Mouse myeloma cell line NS0-derived recombinant human Chitinase 3-like 2 Tyr27-Leu390 Accession # Q15782
100 μg
Primary
Detects human Chitinase 3-like 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human CHIT-1 is observed.
Human
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
CHI3L2, Chitinase 3 like 2, chitinase 3-like 2, chitinase-3-like protein 2, Chondrocyte protein 39, YKL39, YKL-39YKL39
Sheep
Affinity Purified
RUO
1117
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.