missing translation for 'onlineSavingsMsg'
Learn More

Human CCL22/MDC Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 9 publications

£184.00 - £1795.00

Specifications

Antigen CCL22/MDC
Clone 57226
Dilution Western Blot 1 ug/mL, Neutralization 0.6-3.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18794499
GREENER_CHOICE
View Documents
R&D Systems
MAB336-SP
25 μg
£184.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18769967
GREENER_CHOICE
View Documents
R&D Systems
MAB336-100
100 μg
£447.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18798719
GREENER_CHOICE
View Documents
R&D Systems
MAB336-500
500 μg
£1795.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CCL22/MDC Monoclonal specifically detects CCL22/MDC in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

CCL22/MDC
Western Blot 1 ug/mL, Neutralization 0.6-3.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1
CCL22
IgG2b
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
57226
Western Blot, Neutralization, ELISA
Unconjugated
Mouse
Human
O00626.1
6367
E. coli-derived recombinant human CCL22/MDC Gly25-Gln93 Accession # O00626.1
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human CCL22/MDC in ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant mouse CCL17, recombinant human (rh) CCL17, or rhCCL21 is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato