missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL22/MDC Antibody, R&D Systems™

Description
CCL22/MDC Monoclonal specifically detects CCL22/MDC in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
Specifications
| Antigen | CCL22/MDC |
| Applications | Western Blot, Neutralization, ELISA |
| Classification | Monoclonal |
| Clone | 57226 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.6-3.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | O00626.1 |
| Gene Alias | A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 |
| Gene Symbols | CCL22 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?