missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL22/MDC Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems MAB336-500
This item is not returnable.
View return policy
Description
CCL22/MDC Monoclonal specifically detects CCL22/MDC in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CCL22/MDC | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6367 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Neutralization, ELISA | |
| 57226 | |
| Western Blot 1 ug/mL, Neutralization 0.6-3.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| O00626.1 | |
| CCL22 | |
| E. coli-derived recombinant human CCL22/MDC Gly25-Gln93 Accession # O00626.1 | |
| 500 μg | |
| Primary | |
| Detects human CCL22/MDC in ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant mouse CCL17, recombinant human (rh) CCL17, or rhCCL21 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction