missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HP1 Alpha, Beta, and Gamma Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £318.00
Specifications
| Antigen | HP1 Alpha, Beta, and Gamma |
|---|---|
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18389614
|
Novus Biologicals
NBP3-18651-25UL |
25 μg |
£139.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18301064
|
Novus Biologicals
NBP3-18651-100UL |
100 μg |
£318.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HP1 Alpha, Beta, and Gamma Polyclonal antibody specifically detects HP1 Alpha, Beta, and Gamma in Human samples. It is validated for Western BlotSpecifications
| HP1 Alpha, Beta, and Gamma | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P45973 | |
| 23468 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Whole antiserum | |
| Antigen p25, CBXM31HP1-BETA, chromobox homolog 1, chromobox homolog 1 (Drosophila HP1 beta), chromobox homolog 1 (HP1 beta homolog Drosophila ), chromobox homolog 3, chromobox homolog 3 (Drosophila HP1 gamma), chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 1, chromobox protein homolog 3, chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, Heterochromatin protein 1 homolog beta, Heterochromatin protein 1 homolog gamma, heterochromatin protein 1-beta, heterochromatin protein HP1 gamma, Heterochromatin protein p25, heterochromatin protein p25 beta, heterochromatin-like protein 1, HP1, HP1 alpha, HP1 alpha homolog, HP1 beta, HP1 beta homolog, HP1 gamma, HP1A, HP1-ALPHA, HP1-GAMMA, HP1Hs alpha, HP1Hs-alpha, HP1Hsbeta, HP1Hs-beta, MOD1, Modifier 1 protein, Modifier 2 protein, p25beta | |
| Polyclonal antibody raised in rabbit against human HP1 Alpha, Beta, and Gamma (Heterochromatin protein 1 homolog beta), using the full length recombinant GST tagged protein. The antibody also recognizes the alpha and gamma isoforms. | |
| Whole Antiserum |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title