missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HP1 Alpha, Beta, and Gamma Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18651-100UL
This item is not returnable.
View return policy
Description
HP1 Alpha, Beta, and Gamma Polyclonal antibody specifically detects HP1 Alpha, Beta, and Gamma in Human samples. It is validated for Western Blot
Specifications
| HP1 Alpha, Beta, and Gamma | |
| Polyclonal | |
| Western Blot 1:1000 | |
| P45973 | |
| Rabbit | |
| Whole Antiserum | |
| RUO | |
| 23468 | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Whole antiserum | |
| Antigen p25, CBXM31HP1-BETA, chromobox homolog 1, chromobox homolog 1 (Drosophila HP1 beta), chromobox homolog 1 (HP1 beta homolog Drosophila ), chromobox homolog 3, chromobox homolog 3 (Drosophila HP1 gamma), chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 1, chromobox protein homolog 3, chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, Heterochromatin protein 1 homolog beta, Heterochromatin protein 1 homolog gamma, heterochromatin protein 1-beta, heterochromatin protein HP1 gamma, Heterochromatin protein p25, heterochromatin protein p25 beta, heterochromatin-like protein 1, HP1, HP1 alpha, HP1 alpha homolog, HP1 beta, HP1 beta homolog, HP1 gamma, HP1A, HP1-ALPHA, HP1-GAMMA, HP1Hs alpha, HP1Hs-alpha, HP1Hsbeta, HP1Hs-beta, MOD1, Modifier 1 protein, Modifier 2 protein, p25beta | |
| Polyclonal antibody raised in rabbit against human HP1 Alpha, Beta, and Gamma (Heterochromatin protein 1 homolog beta), using the full length recombinant GST tagged protein. The antibody also recognizes the alpha and gamma isoforms. | |
| 100 μg | |
| Primary | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction