missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HP1 Alpha, Beta, and Gamma Rabbit anti-Human, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
HP1 Alpha, Beta, and Gamma Polyclonal antibody specifically detects HP1 Alpha, Beta, and Gamma in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | HP1 Alpha, Beta, and Gamma |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000 |
| Formulation | Whole antiserum |
| Gene Accession No. | P45973 |
| Gene Alias | Antigen p25, CBXM31HP1-BETA, chromobox homolog 1, chromobox homolog 1 (Drosophila HP1 beta), chromobox homolog 1 (HP1 beta homolog Drosophila ), chromobox homolog 3, chromobox homolog 3 (Drosophila HP1 gamma), chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 1, chromobox protein homolog 3, chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, Heterochromatin protein 1 homolog beta, Heterochromatin protein 1 homolog gamma, heterochromatin protein 1-beta, heterochromatin protein HP1 gamma, Heterochromatin protein p25, heterochromatin protein p25 beta, heterochromatin-like protein 1, HP1, HP1 alpha, HP1 alpha homolog, HP1 beta, HP1 beta homolog, HP1 gamma, HP1A, HP1-ALPHA, HP1-GAMMA, HP1Hs alpha, HP1Hs-alpha, HP1Hsbeta, HP1Hs-beta, MOD1, Modifier 1 protein, Modifier 2 protein, p25beta |
| Host Species | Rabbit |
| Immunogen | Polyclonal antibody raised in rabbit against human HP1 Alpha, Beta, and Gamma (Heterochromatin protein 1 homolog beta), using the full length recombinant GST tagged protein. The antibody also recognizes the alpha and gamma isoforms. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?