missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HP1 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£165.00 - £391.00
Specifications
| Antigen | HP1 alpha |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:500, Flow (Intracellular) 1:500 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18074284
|
Novus Biologicals
NB100-2518SS |
0.025 mL |
£165.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18018633
|
Novus Biologicals
NB100-2518 |
0.1 mL |
£391.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HP1 alpha Polyclonal specifically detects HP1 alpha in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).Specifications
| HP1 alpha | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS with 0.02% Sodium Azide | |
| Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha | |
| CBX5 | |
| IgG | |
| IgG purified | |
| Human. Expected to react with mouse, Arabidopsis suecica, Rye and Triticum aestivum due to sequence homology. |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:500, Flow (Intracellular) 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| P45973 | |
| 23468 | |
| A synthetic peptide containing an N-terminal region of human HP1 alpha (within residues 1-100). [Swiss-Prot P45973] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 20 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title