missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HP1 alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2518SS
This item is not returnable.
View return policy
Description
HP1 alpha Polyclonal specifically detects HP1 alpha in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).
Specifications
| HP1 alpha | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:500, Flow (Intracellular) 1:500 | |
| P45973 | |
| CBX5 | |
| A synthetic peptide containing an N-terminal region of human HP1 alpha (within residues 1-100). [Swiss-Prot P45973] | |
| IgG purified | |
| RUO | |
| Primary | |
| Human. Expected to react with mouse, Arabidopsis suecica, Rye and Triticum aestivum due to sequence homology. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha | |
| Rabbit | |
| 20 kDa | |
| 0.025 mL | |
| Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 23468 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction