missing translation for 'onlineSavingsMsg'
Learn More

HP1 alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-2518

 View more versions of this product

Product Code. 18018633

  • £391.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HP1 alpha Polyclonal specifically detects HP1 alpha in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HP1 alpha
Polyclonal
Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:500, Flow (Intracellular) 1:500
P45973
CBX5
A synthetic peptide containing an N-terminal region of human HP1 alpha (within residues 1-100). [Swiss-Prot P45973]
IgG purified
RUO
Primary
Human. Expected to react with mouse, Arabidopsis suecica, Rye and Triticum aestivum due to sequence homology.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry
Unconjugated
PBS with 0.02% Sodium Azide
Antigen p25, chromobox homolog 5, chromobox homolog 5 (Drosophila HP1 alpha), chromobox homolog 5 (HP1 alpha homolog, Drosophila), chromobox protein homolog 5, Heterochromatin protein 1 homolog alpha, heterochromatin protein 1-alpha, HP1, HP1 alpha, HP1 alpha homolog, HP1A, HP1-ALPHA, HP1Hs alpha, HP1Hs-alpha
Rabbit
20 kDa
0.1 mL
Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing
23468
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only