missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap IXSelect
HiTrap IXSelect columns are prepacked with IXSelect resin for screening and small-scale protein purifications using affinity chromatography.
£815.00 - £817.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15579261
|
Cytiva
17371411 |
5 x 1 mL |
£817.00
Pack of 5 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
15589261
|
Cytiva
17371412 |
1 x 5 mL |
£815.00
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
- XSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams.
- Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
Specifications
2°C to 8°C |