missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap IXSelect

HiTrap IXSelect columns are prepacked with IXSelect resin for screening and small-scale protein purifications using affinity chromatography.

Brand:  Cytiva 17371412

Additional Details : Weight : 0.08000kg


 View more versions of this product

Product Code. 15589261

  • £815.00 / Each
Estimated Shipment: 09-07-2024
to see stock.
Add to basket

Description

Description

  • XSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams.
  • Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications.
Specifications

Specifications

2°C to 8°C
1 x 5 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap IX select, 5 ml, 1 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers