Learn More
Cytiva HisTrap™ FF Crude Prepacked Columns
Purify histidine-tagged proteins directly from homogenized, unclarified cell lysates. Cytiva Life Sciences™ HisTrap™ FF Crude Prepacked Columns offer the convenience of simple, one-step purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).
Brand: Cytiva 17-5286-02
Code : GX
Additional Details : CAS Number : 79-06-1 Weight : 1.52000kg
Description
- Prepacked with Ni Sepharose™ 6 Fast Flow media
- Binding capacity approximately 40mg/mL medium with negligible Ni2+ leakage
- Maximum pressure: 3 bar
- Column material: polypropylene
- Compatible with wide variety of reducing agents, detergents, denaturants, and other additives
- 1 and 5mL columns
Media
- Matrix: 6% cross-linked agarose
- Average particle size: 90μm
- pH stability working range: 3 to 12
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
5 mL | |
16 mm | |
Polypropylene | |
6% Cross-linked Agarose | |
3 bar | |
100 x 5 mL |
0.6 in. | |
<20 mL/min. | |
3 to 12 | |
4°C to 30°C | |
45 to 165 μm | |
Pre-packed Column |
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol, Nickel