missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Sepharose™ Prepacked High Performance HIC Columns
Designed for method optimization and parameter screening
£139.00 - £140.00
Specifications
Bed Volume | 4.7mL |
---|---|
Diameter (English) Inner | 0.3 in. |
Diameter (Metric) Inner | 7.7mm |
Quantity | 1 x 4.7mL |
Height (English) | 3.93 in. |
Product Code | Brand | Buffer | Product Type | Ligand Type | Support Type | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Buffer | Product Type | Ligand Type | Support Type | Price | Quantity & Availability | ||
12475700
|
Cytiva
28-9782-42 |
1M NaOH, 1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Aceto nitrile, 70% Ethanol, 3M Ammoniumsulfate, 1mM HCl, 30% Isopropanol, 2% SDS | HiScreen Butyl Sepharose HP column | Butyl | Cross-linked Agarose |
£139.00
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||
10557680
|
Cytiva
28-9505-16 |
1M NaOH, 1M Acetic Acid, 30% Aceto nitrile, 70% Ethanol, 30% Isopropanol, 6M Guanidine Hydrochloride, 8M Ure, 3M Ammoniumsulfate, Ionic Detergents, Non ionic Detergents, Polar Organic Solvents, 2% Sodium Dodectyl Sulfate | HiScreen Phenyl Sepharose HP column | Phenyl | 6% Cross-linked Agarose |
£140.00
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||
Description
- Ready-to-use columns for convenient process development optimization
- Standard aliphatic/aromatic Hydrophobic Interaction Chromatography media for capture and intermediate purification
- Bed height of 10cm designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20cm bed height
- Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
HiScreen Phenyl HP
- Degree of substitution: ∽25μmol phenyl/mL gel
- Degree of substitution: ∽50μmol phenyl/mL gel
Specifications
4.7mL | |
7.7mm | |
3.93 in. | |
<150cm/hr. | |
3 bar | |
24 to 44μm | |
Ion Exchange Column |
0.3 in. | |
1 x 4.7mL | |
100mm | |
Polypropylene | |
3 to 13 | |
4° to 30°C | |
Convenient process development optimization parameter screening |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiScreen Butyl HP
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.