missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Sepharose™ Prepacked High Performance HIC Columns
Shop All Cytiva Products
Click to view available options
Ligand Type:
Butyl
Phenyl
Unit Size:
Each
Description
- Ready-to-use columns for convenient process development optimization
- Standard aliphatic/aromatic Hydrophobic Interaction Chromatography media for capture and intermediate purification
- Bed height of 10cm designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20cm bed height
- Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
- Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
HiScreen Phenyl HP
- Degree of substitution: ∽25μmol phenyl/mL gel
- Degree of substitution: ∽50μmol phenyl/mL gel
Specifications
Specifications
| Buffer | 1M NaOH, 1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Aceto nitrile, 70% Ethanol, 3M Ammoniumsulfate, 1mM HCl, 30% Isopropanol, 2% SDS |
| Product Type | HiScreen Butyl Sepharose HP column |
| Bed Volume | 4.7mL |
| Diameter (English) Inner | 0.3 in. |
| Diameter (Metric) Inner | 7.7mm |
| Quantity | 1 x 4.7 mL |
| Height (English) | 3.93 in. |
| Height (Metric) | 100mm |
| Flow Rate | <150 cm/hr. |
| Material | Polypropylene |
| Show More |
Safety and Handling
Product Identifier
- HiScreen Butyl HP
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction