missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation High/Low pH UPLC Method Development Kit
The High/Low pH UPLC method development kit combines BEH C18, C8, Shield RP18 and Phenyl column chemistries to accomodate your method development approach, enabling methods to be developed efficiently and effectively.
£4290.00 - £4560.00
Specifications
Max. Pressure | 18000 psi (1240 Bar) |
---|---|
Particle Shape | Spherical |
Particle Size | 1.7 μm |
Pore Size | 130 Å |
Quantity | 4/Pk. |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
10135674
|
Waters Corporation
176001042 |
2.1 mm | 50 mm |
£4290.00
Pack of 4 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10753234
|
Waters Corporation
176001043 |
2.1 mm | 100 mm |
£4560.00
Pack of 4 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
13270030
|
Waters Corporation
176001881 |
3 mm | 50 mm |
£4290.00
Pack of 4 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
13280030
|
Waters Corporation
176001882 |
3 mm | 100 mm |
£4560.00
Pack of 4 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
Description
- By utilizing UPLC Technology for method development, a substantial improvement in throughput can be realized.
- This, in turn, reduces cost per sample and time of analysis considerably while maintaining or improving separation integrity, enabling products to be brought to market faster and improving the overall profitability of the assay.
Specifications
18000 psi (1240 Bar) | |
1.7 μm | |
4/Pk. | |
Method Development Kit |
Spherical | |
130 Å | |
Yes |