missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation High/Low pH UPLC Method Development Kit

The High/Low pH UPLC method development kit combines BEH C18, C8, Shield RP18 and Phenyl column chemistries to accomodate your method development approach, enabling methods to be developed efficiently and effectively.

Brand:  Waters Corporation 176001882

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13280030

  • £4560.00 / Pack of 4
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

  • By utilizing UPLC Technology for method development, a substantial improvement in throughput can be realized.
  • This, in turn, reduces cost per sample and time of analysis considerably while maintaining or improving separation integrity, enabling products to be brought to market faster and improving the overall profitability of the assay.
Specifications

Specifications

3 mm
18000 psi (1240 Bar)
1.7 μm
4/Pk.
Method Development Kit
100 mm
Spherical
130 Å
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers