missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation High/Low pH UPLC Method Development Kit

Product Code. 10135674
Encompass
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
Diameter (Metric):
2.1 mm
3 mm
Unit Size:
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
10135674 50 mm 2.1 mm Pack of 4
10753234 100 mm 2.1 mm Pack of 4
13270030 50 mm 3 mm Pack of 4
13280030 100 mm 3 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10135674 Supplier Waters Corporation Supplier No. 176001042

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The High/Low pH UPLC method development kit combines BEH C18, C8, Shield RP18 and Phenyl column chemistries to accomodate your method development approach, enabling methods to be developed efficiently and effectively.

  • By utilizing UPLC Technology for method development, a substantial improvement in throughput can be realized.
  • This, in turn, reduces cost per sample and time of analysis considerably while maintaining or improving separation integrity, enabling products to be brought to market faster and improving the overall profitability of the assay.

Specifications

Diameter (Metric) 2.1 mm
Length (Metric) 50 mm
Max. Pressure 18000 psi (1240 Bar)
Particle Shape Spherical
Particle Size 1.7 μm
Pore Size 130 Å
Quantity 4/Pk.
Endcapped Yes
Type Method Development Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.