Learn More
Heptanal, 97%, Thermo Scientific Chemicals
£16.50 - £65.20
Chemical Identifiers
CAS | 111-71-7 |
---|---|
Molecular Formula | C7H14O |
Molecular Weight (g/mol) | 114.19 |
MDL Number | MFCD00007028 |
InChI Key | FXHGMKSSBGDXIY-UHFFFAOYSA-N |
Synonym | heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal |
PubChem CID | 8130 |
ChEBI | CHEBI:34787 |
IUPAC Name | heptanal |
SMILES | CCCCCCC=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11389819
|
Thermo Scientific Alfa Aesar
B23830.AE |
100 mL |
£16.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369819
|
Thermo Scientific Alfa Aesar
B23830.AK |
250 mL |
£24.80
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399819
|
Thermo Scientific Alfa Aesar
B23830.AP |
500 mL |
£37.90
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11379819
|
Thermo Scientific Alfa Aesar
B23830.AU |
1000 mL |
£65.20
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHeptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.
Solubility
Soluble in water 1.3 g/L.
Notes
Hygroscopic. Store away from oxidizing agents, air.
Chemical Identifiers
111-71-7 | |
114.19 | |
FXHGMKSSBGDXIY-UHFFFAOYSA-N | |
8130 | |
heptanal |
C7H14O | |
MFCD00007028 | |
heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal | |
CHEBI:34787 | |
CCCCCCC=O |
Specifications
111-71-7 | |
0.85 | |
35°C (95°F) | |
Unpleasant | |
1.4125 | |
UN3056 | |
Air Sensitive | |
heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal | |
FXHGMKSSBGDXIY-UHFFFAOYSA-N | |
heptanal | |
8130 | |
114.19 | |
Liquid |
-43°C | |
152°C to 154°C | |
0.97 | |
C7H14O | |
MFCD00007028 | |
1560236 | |
14,4658 | |
Soluble in water 1.3g/L. | |
CCCCCCC=O | |
114.19 | |
CHEBI:34787 | |
97% | |
Heptanal |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H226-H315
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: N-HEPTALDEHYDE
missing translation for 'einecsNumber' : 203-898-4
missing translation for 'rtecsNumber' : MI6900000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only