Learn More
Heptanal, 97%
Description
Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.
Solubility
Soluble in water 1.3 g/L.
Notes
Hygroscopic. Store away from oxidizing agents, air.
Specifications
Specifications
| Melting Point | -43°C |
| Density | 0.85 |
| Boiling Point | 152°C to 154°C |
| Flash Point | 35°C (95°F) |
| Assay Percent Range | 0.97 |
| Odor | Unpleasant |
| Refractive Index | 1.4125 |
| Quantity | 100 mL |
| UN Number | UN3056 |
| Beilstein | 1560236 |
| Show More |
Safety and Handling
- Heptanal
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- MIXTURE LIST-Contain: Heptanal
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.