missing translation for 'onlineSavingsMsg'
Learn More

Heptanal, 97%

Product Code. 11379819
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1000 mL
Unit Size:
1000mL
100mL
250mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379819 1000 mL 1000mL
11389819 100 mL 100mL
11369819 250 mL 250mL
11399819 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11379819 Supplier Thermo Scientific Alfa Aesar Supplier No. B23830.AU
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-71-7 | C7H14O | 114.19 g/mol

Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptanal is used as a synthesis intermediate in the fragrances and flavors industry. It is the precursor to 1-heptanol, ethyl heptanoate and for certain lubricants.

Solubility
Soluble in water 1.3 g/L.

Notes
Hygroscopic. Store away from oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-71-7
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00007028
InChI Key FXHGMKSSBGDXIY-UHFFFAOYSA-N
Synonym heptaldehyde, enanthaldehyde, n-heptaldehyde, enanthal, heptyl aldehyde, heptanaldehyde, n-heptanal, oenanthaldehyde, enanthole, oenanthal
PubChem CID 8130
ChEBI CHEBI:34787
IUPAC Name heptanal
SMILES CCCCCCC=O

Specifications

Melting Point -43°C
Density 0.85
Boiling Point 152°C to 154°C
Flash Point 35°C (95°F)
Assay Percent Range 0.97
Odor Unpleasant
Refractive Index 1.4125
Quantity 1000 mL
UN Number UN3056
Beilstein 1560236
Sensitivity Air Sensitive
Merck Index 14,4658
Solubility Information Soluble in water 1.3g/L.
Formula Weight 114.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Heptanal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Heptanal
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Heptanal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.