Learn More
Hepatocyte Growth Factor Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results with Epredia™ Hepatocyte Growth Factor Ab-1, Mouse Monoclonal Antibody.
£105.06 - £483.48
Specifications
Antigen | Hepatocyte Growth Factor Ab-1 |
---|---|
Clone | 4C12.1 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Concentrated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12611927
|
Epredia
MS-651-P0 |
200μg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12621927
|
Epredia
MS-651-P1 |
200μg/mL | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12601927
|
Epredia
MS-651-P |
200μg/mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12631927
|
Epredia
MS-651-P1ABX |
1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12641927
|
Epredia
MS-651-PABX |
1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Hepatocyte Growth Factor (HGF), also known as Scatter Factor (SF) or Hepatopoietin A. Membrane receptor for HGF/SF is the product of the proto-oncogene c-met, a tyrosine kinase. HGF/SF is a pleiotropic growth factor which is produced by mesodermally derived cells, such as Kupffer cells/macrophages, endothelial cells, or hepatic fat-storing cells.Host Species: Mouse
Clone: 4C12.1
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human HGF protein
Molecular Weight: ∽82kDa
Recommended for:
- Western Blotting (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Hepatocyte Growth Factor Ab-1 | |
Monoclonal | |
Concentrated | |
RUO | |
Human | |
IgG2b |
4C12.1 | |
Unconjugated | |
Mouse | |
Growth Factor | |
Recombinant human HGF protein | |
Primary |