Learn More
Hepatocyte Growth Factor Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results with Epredia™ Hepatocyte Growth Factor Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-651-P0
Code : LD
Additional Details : Weight : 1.00000kg
Description
Hepatocyte Growth Factor (HGF), also known as Scatter Factor (SF) or Hepatopoietin A. Membrane receptor for HGF/SF is the product of the proto-oncogene c-met, a tyrosine kinase. HGF/SF is a pleiotropic growth factor which is produced by mesodermally derived cells, such as Kupffer cells/macrophages, endothelial cells, or hepatic fat-storing cells.Host Species: Mouse
Clone: 4C12.1
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human HGF protein
Molecular Weight: ∽82kDa
Recommended for:
- Western Blotting (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Hepatocyte Growth Factor Ab-1 | |
4C12.1 | |
Unconjugated | |
Recombinant human HGF protein | |
RUO | |
Primary | |
Concentrated |
Monoclonal | |
200μg/mL | |
Mouse | |
100 μL | |
Growth Factor | |
Human | |
IgG2b |