missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7, p Ser155 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£171.00 - £318.00
Specifications
| Antigen | HDAC7 (p Ser155) |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.1-1 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18120722
|
Novus Biologicals
NBP2-24454SS |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18157201
|
Novus Biologicals
NBP2-24454 |
0.05 mg |
£318.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC7 Polyclonal specifically detects HDAC7 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.Specifications
| HDAC7 (p Ser155) | |
| Western Blot 0.1-1 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Chromatin Research, Phospho Specific | |
| 0.1 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 51564 | |
| A synthetic peptide containing phospho-serine 155 of human HDAC-7 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Bovine, Canine, Primate | |
| DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 | |
| HDAC7 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, and dog, 94% homologous in chicken, opossum and pufferfish and 83% homologous in zebrafish. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title