missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7, p Ser155 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24454SS
This item is not returnable.
View return policy
Description
HDAC7 Polyclonal specifically detects HDAC7 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.
Specifications
| HDAC7 (p Ser155) | |
| Polyclonal | |
| Unconjugated | |
| 0.1 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| HDAC7 | |
| A synthetic peptide containing phospho-serine 155 of human HDAC-7 was used as the immunogen. | |
| 0.025 mg | |
| Breast Cancer, Cancer, Chromatin Research, Phospho Specific | |
| 51564 | |
| Human, Mouse, Rat, Bovine, Canine, Primate | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.1-1 ug/ml | |
| DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, and dog, 94% homologous in chicken, opossum and pufferfish and 83% homologous in zebrafish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction