missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7, p Ser155 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NBP2-24454
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
HDAC7 Polyclonal specifically detects HDAC7 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.
Spezifikation
| HDAC7 (p Ser155) | |
| Polyclonal | |
| Unconjugated | |
| 0.1 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| HDAC7 | |
| A synthetic peptide containing phospho-serine 155 of human HDAC-7 was used as the immunogen. | |
| 0.05 mg | |
| Breast Cancer, Cancer, Chromatin Research, Phospho Specific | |
| 51564 | |
| Human, Mouse, Rat, Bovine, Canine, Primate | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.1-1 ug/ml | |
| DKFZP586J0917, EC 3.5.1.98, HD7, HD7A, HDAC7AFLJ99588, histone deacetylase 7, Histone deacetylase 7ADKFZp586J0917 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, and dog, 94% homologous in chicken, opossum and pufferfish and 83% homologous in zebrafish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur