missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haemophilus Influenzae B Antibody (1079/457) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£160.00 - £397.00
Specifications
| Antigen | Haemophilus Influenzae B |
|---|---|
| Clone | 1079/457 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Immunofluorescence, Immunocytochemistry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18063496
|
Novus Biologicals
NB100-64551-0.025MG |
0.025 mg |
£160.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18016295
|
Novus Biologicals
NB100-64551 |
0.1 mg |
£397.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Haemophilus Influenzae B Monoclonal specifically detects Haemophilus Influenzae B in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Haemophilus Influenzae B | |
| 1.0 mg/mL | |
| Immunofluorescence, Immunocytochemistry, ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| Native, NCTC 7279 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1079/457 | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| H. influenza B | |
| IgG2a | |
| Protein G purified | |
| NB100-64551 recognizes Haemophilus influenzae B, a gram-negative coccobacillus that is the cause of bacterial meningitis and other severe illnesses in young children. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title