missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haemophilus Influenzae B Antibody (1079/457) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64551-0.025MG
This item is not returnable.
View return policy
Description
Haemophilus Influenzae B Monoclonal specifically detects Haemophilus Influenzae B in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Spécification
| Haemophilus Influenzae B | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| H. influenza B | |
| Native, NCTC 7279 | |
| 0.025 mg | |
| Primary | |
| Bacteria | |
| Purified |
| Immunofluorescence, Immunocytochemistry, ELISA | |
| 1079/457 | |
| Unconjugated | |
| PBS (pH 7.2) with 0.09% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| NB100-64551 recognizes Haemophilus influenzae B, a gram-negative coccobacillus that is the cause of bacterial meningitis and other severe illnesses in young children. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu