missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haemophilus Influenzae B Antibody (1079/457) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64551
This item is not returnable.
View return policy
Description
Haemophilus Influenzae B Monoclonal specifically detects Haemophilus Influenzae B in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Haemophilus Influenzae B | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| NB100-64551 recognizes Haemophilus influenzae B, a gram-negative coccobacillus that is the cause of bacterial meningitis and other severe illnesses in young children. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| 1079/457 | |
| Unconjugated | |
| H. influenza B | |
| Native, NCTC 7279 | |
| 0.1 mg | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction