missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme B Antibody (GB11), Novus Biologicals™
Mouse Monoclonal Antibody
£175.00 - £533.00
Specifications
| Antigen | Granzyme B |
|---|---|
| Clone | GB11 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18092004
|
Novus Biologicals
NB100-64774-0.025MG |
0.025 mg |
£175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18032866
|
Novus Biologicals
NB100-64774-0.05MG |
0.05 mg |
£378.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18091344
|
Novus Biologicals
NB100-64774 |
0.1 mg |
£533.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Granzyme B Monoclonal specifically detects Granzyme B in Human, Primate, Monkey samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| Granzyme B | |
| 1 mg/mL | |
| Monoclonal | |
| Mouse | |
| P10144 | |
| GZMB | |
| IgG1 | |
| NB100-64774 recognizes the serine protease Granzyme B (GZMB) |
| GB11 | |
| Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, ELISA | |
| Unconjugated | |
| Primate, Monkey, Human | |
| 3002 | |
| Purified human Granzyme B (GZMB) | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title