missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme B Antibody (GB11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64774
This item is not returnable.
View return policy
Beskrivelse
Granzyme B Monoclonal specifically detects Granzyme B in Human, Primate, Monkey samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
Tekniske data
| Granzyme B | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10-1:100, ELISA 2 ug/ml-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GranzymeB, GZMB, HLP, SECT | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3002 | |
| Human, Primate, Monkey | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| GB11 | |
| Unconjugated | |
| P10144 | |
| GZMB | |
| Purified human Granzyme B (GZMB) | |
| 0.1 mg | |
| Primary | |
| NB100-64774 recognizes the serine protease Granzyme B (GZMB) | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion