missing translation for 'onlineSavingsMsg'
Learn More

Granzyme B Antibody (GB11), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB100-64774-0.025MG

 View more versions of this product

Product Code. 18092004

  • £175.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Granzyme B Monoclonal specifically detects Granzyme B in Human, Primate, Monkey samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Granzyme B
Monoclonal
1 mg/mL
Flow Cytometry 1:10-1:100, ELISA 2 ug/ml-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
P10144
GZMB
Purified human Granzyme B (GZMB)
0.025 mg
Primary
NB100-64774 recognizes the serine protease Granzyme B (GZMB)
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, ELISA
GB11
Unconjugated
Tris buffered saline with 0.09% Sodium Azide
C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTSGL1, granzyme B, GZMB, HLP, SECT
Mouse
Protein A purified
RUO
3002
Primate, Monkey, Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only