missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Very thin paper
- Made with 100% Manilla fibers
- Not fluffy
- Weight: 12g/m2
Application
- Cleaning of various optical lens
- Cleaning of optical fibers filaments
Specifications
Specifications
| Product Type | Lens Cleaning Tissue |
| For Use With (Application) | Cleaning of various optical lens (objective, microscopes binoculars, magnifying Glass, Glasses, etc.), cleaning of optical fibers filaments |
| Material | Manilla Fibers |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?