missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOSR1 Antibody (HFD9), Novus Biologicals™
Mouse Monoclonal Antibody
£145.00 - £633.00
Specifications
| Antigen | GOSR1 |
|---|---|
| Clone | HFD9 |
| Dilution | Western Blot 1:1000/ECL, Electron Microscopy 1:10-1:500 |
| Applications | Western Blot, Immunoassay |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18069928
|
Novus Biologicals
NBP1-97437-0.025MG |
0.025 mg |
£145.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18064818
|
Novus Biologicals
NBP1-97437-0.05MG |
0.05 mg |
£427.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18017729
|
Novus Biologicals
NBP1-97437-0.2MG |
0.2 mg |
£633.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GOSR1 Monoclonal specifically detects GOSR1 in Mouse, Rat, Porcine, Bovine, Canine, Chicken, Rabbit, Sheep samples. It is validated for Western Blot, Electron Microscopy.Specifications
| GOSR1 | |
| Western Blot 1:1000/ECL, Electron Microscopy 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Bovine, Rabbit, Pig, Sheep, Chicken, Canine, Rat | |
| golgi integral membrane protein 2, golgi SNAP receptor complex member 1,28 kDa cis-Golgi SNARE p28, Golgi SNARE 28 kDa, GOLIM2, GOS28cis-golgi SNARE, GOS-28GOS28/P28, GS2828 kDa Golgi SNARE protein, P28 | |
| GOSR1 | |
| IgG1 | |
| Protein G purified |
| HFD9 | |
| Western Blot, Immunoassay | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 9527 | |
| Rat liver Golgi-enriched membranes. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title