missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOSR1 Antibody (HFD9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97437-0.05MG
This item is not returnable.
View return policy
Description
GOSR1 Monoclonal specifically detects GOSR1 in Mouse, Rat, Porcine, Bovine, Canine, Chicken, Rabbit, Sheep samples. It is validated for Western Blot, Electron Microscopy.
Specifications
| GOSR1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| GOSR1 | |
| Rat liver Golgi-enriched membranes. | |
| 0.05 mg | |
| Membrane Trafficking and Chaperones | |
| 9527 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunoassay | |
| HFD9 | |
| Western Blot 1:1000/ECL, Electron Microscopy 1:10-1:500 | |
| golgi integral membrane protein 2, golgi SNAP receptor complex member 1,28 kDa cis-Golgi SNARE p28, Golgi SNARE 28 kDa, GOLIM2, GOS28cis-golgi SNARE, GOS-28GOS28/P28, GS2828 kDa Golgi SNARE protein, P28 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, Rat, Pig, Bovine, Canine, Chicken, Rabbit, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction