missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GnRH1 Monoclonal Antibody (LHRH13-426.12), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£367.00 - £916.00

Specifications

Antigen GnRH1
Clone LHRH13-426.12
Concentration 1.66 mg/mL
Applications ELISA, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17890466
View Documents
Invitrogen™
MA5-46100
100 μg
£367.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11593423
GREENER_CHOICE
View Documents
Invitrogen™
MA110817
500 μg
£480.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17800186
View Documents
Invitrogen™
MA5-46179
1 mg
£916.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MA1-10817 detects Gonadotropin Releasing Hormone from human samples. MA1-10817 has been successfully used in ELISA, radioimmune assays and immunohistochemistry applications. The MA1-10817 immunogen is purified human gonadotropin releasing hormone.

The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).
Specifications

Specifications

GnRH1
1.66 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.4
Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1
GNRH1
IgG1
Protein A
Antibody
LHRH13-426.12
ELISA, Immunohistochemistry, Radioimmune Assays (RIA)
Unconjugated
Mouse
Human
P01148
2796
Purified human gonadotropin releasing hormone.
Primary
-20°C, Avoid Freeze/Thaw Cycles
GNRH1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.