Learn More
Invitrogen™ GnRH1 Monoclonal Antibody (LHRH13-426.12), Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA5-46100
 View more versions of this product
Product Code. 17890466
  • £367.00 / 100µg
Description
MA1-10817 detects Gonadotropin Releasing Hormone from human samples. MA1-10817 has been successfully used in ELISA, radioimmune assays and immunohistochemistry applications. The MA1-10817 immunogen is purified human gonadotropin releasing hormone.
The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GnRH1
Monoclonal
1.66 mg/mL
PBS with no preservative; pH 7.4
P01148
GNRH1
Purified human gonadotropin releasing hormone.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Immunohistochemistry, Radioimmune Assays (RIA)
LHRH13-426.12
Unconjugated
GNRH1
Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1
Mouse
Protein A
RUO
2796
-20°C, Avoid Freeze/Thaw Cycles