missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GnRH1 Monoclonal Antibody (LHRH13-426.12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA5-46179
This item is not returnable.
View return policy
Description
MA1-10817 detects Gonadotropin Releasing Hormone from human samples. MA1-10817 has been successfully used in ELISA, radioimmune assays and immunohistochemistry applications. The MA1-10817 immunogen is purified human gonadotropin releasing hormone.
The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).
Specifications
| GnRH1 | |
| Monoclonal | |
| 1.66 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P01148 | |
| GNRH1 | |
| Purified human gonadotropin releasing hormone. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry, Radioimmune Assays (RIA) | |
| LHRH13-426.12 | |
| Unconjugated | |
| GNRH1 | |
| Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2796 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction