missing translation for 'onlineSavingsMsg'
Learn More

Glycophorin A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen Glycophorin A
Concentration 0.33 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16895254
View Documents
Proteintech
15874-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16885254
View Documents
Proteintech
15874-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD235a (Glycophorin A, GPA) is a transmembrane sialoglycoprotein expressed on erythrocytes and their precursors. Similarly to glycophorin B (GPB), these molecules provide the cells with a large mucin-like surface, which minimalizes aggregation between erythrocytes in the circulation. GPA is the carrier of blood group M and N specificities, while GPB accounts for S, s and U specificities. CD235a is a receptor of Hsa, an Streptococcus adhesin.
Specifications

Specifications

Glycophorin A
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry
Unconjugated
Rabbit
Human
P02724
2993
Glycophorin A Fusion Protein Ag8635
Primary
-20°C
GYPA
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CD235a, Glycophorin A, GPA, GPErik, GpMiIII, GPSAT, GYPA, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), MN, MN sialoglycoprotein, MNS, PAS 2, Sialoglycoprotein alpha
GYPA
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.