missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycophorin A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15874-1-AP-150UL
This item is not returnable.
View return policy
Description
CD235a (Glycophorin A, GPA) is a transmembrane sialoglycoprotein expressed on erythrocytes and their precursors. Similarly to glycophorin B (GPB), these molecules provide the cells with a large mucin-like surface, which minimalizes aggregation between erythrocytes in the circulation. GPA is the carrier of blood group M and N specificities, while GPB accounts for S, s and U specificities. CD235a is a receptor of Hsa, an Streptococcus adhesin.Specifications
| Glycophorin A | |
| Polyclonal | |
| Unconjugated | |
| GYPA | |
| CD235a, Glycophorin A, GPA, GPErik, GpMiIII, GPSAT, GYPA, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), MN, MN sialoglycoprotein, MNS, PAS 2, Sialoglycoprotein alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2993 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02724 | |
| GYPA | |
| Glycophorin A Fusion Protein Ag8635 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction