Learn More
Glycophorin A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15874-1-AP-20UL
 View more versions of this product
Product Code. 16895254
  • £135.00 / 20µL
Description
CD235a (Glycophorin A, GPA) is a transmembrane sialoglycoprotein expressed on erythrocytes and their precursors. Similarly to glycophorin B (GPB), these molecules provide the cells with a large mucin-like surface, which minimalizes aggregation between erythrocytes in the circulation. GPA is the carrier of blood group M and N specificities, while GPB accounts for S, s and U specificities. CD235a is a receptor of Hsa, an Streptococcus adhesin.
Specifications
Specifications
Glycophorin A
Polyclonal
Unconjugated
GYPA
CD235a, Glycophorin A, GPA, GPErik, GpMiIII, GPSAT, GYPA, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), MN, MN sialoglycoprotein, MNS, PAS 2, Sialoglycoprotein alpha
Rabbit
Antigen Affinity Chromatography
RUO
2993
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02724
GYPA
Glycophorin A Fusion Protein Ag8635
20 μL