missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ IL9 Recombinant Human Protein
Recombinant Human Interleukin 9 (IL9) is a bioactive protein intended for use in cell culture applications
£258.00 - £2155.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 48 kDa |
Source | Human |
Gene Alias | IL-9 |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12053589
|
Gibco™
PHC0094 |
10μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12043589
|
Gibco™
PHC0091 |
100μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
IL9 is produced by CD4+ helper cells, and has been shown to be a pleiotropic cytokine impacting lymphoid and mast cell lineages.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to get a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose-dependent stimulation of the proliferation of human MO7e cells
- Proven compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Dry Ice.
Specifications
Cell Culture | |
Human | |
Human | |
Store in freezer (-5 to -30°C). | |
0.2 ng/ml | |
< 1 EU/ml | |
Mammalian | |
Recombinant, Ligand | |
Interleukins | |
95 % |
48 kDa | |
IL-9 | |
Recombinant | |
Carrier-Free | |
Proliferation of human MO7e cells | |
Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Apoptosis, Stem Cell Research, Oncology, Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.