missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL9 Recombinant Human Protein

Recombinant Human Interleukin 9 (IL9) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC0091

Code : 50

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 12043589

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

IL9 is produced by CD4+ helper cells, and has been shown to be a pleiotropic cytokine impacting lymphoid and mast cell lineages.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to get a biologically active protein
  • High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by the dose-dependent stimulation of the proliferation of human MO7e cells
  • Proven compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Dry Ice.

Specifications

Specifications

Cell Culture
100μg
IL-9
Recombinant
Carrier-Free
Proliferation of human MO7e cells
100 μg
Mammalian
Recombinant, Ligand
Interleukins
95 %
48 kDa
Human
Human
Store in freezer (-5 to -30°C).
0.2 ng/ml
< 1 EU/ml
Dry Ice
Cytokines & Receptors
Lyophilized
Apoptosis, Stem Cell Research, Oncology, Signal Transduction
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.