missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, R&D Systems™

Mouse Monoclonal Antibody
£203.00 - £1590.00
Specifications
| Antigen | GFP |
|---|---|
| Clone | 454505R |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722819
![]() |
R&D Systems
MAB42401R-SP |
25 μg |
£203.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702688
![]() |
R&D Systems
MAB42401R-100 |
100 μg |
£393.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18109620
|
R&D Systems
MAB42401R-01M |
1 mg |
£1460.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18169170
|
R&D Systems
MAB42401R-500 |
500 ug |
£1590.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GFP Monoclonal specifically detects GFP in Multi-Species samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
| GFP | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Multi-species | |
| eGFP, Enhanced Green Fluorescent Protein, GFPuv, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 454505R | |
| Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| P42212 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Stains eGFP transfectants but not irrelevant transfectants in flow cytometry. Detects recombinant GFPuv in Western blots. For optimal Western blot results, we recommend R&D Systems Goat Anti-GFP Antigen Affinity-purified Polyclonal Antibody (Catalog # AF4240 ) or Novus Biologicals Mouse Anti-GFP Monoclonal Antibody (Catalog # NBP2-2211 ). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title