Learn More
GFP Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB42401R-SP
 View more versions of this product
Product Code. 18722819
  • £203.00 / 25µg
Description
GFP Monoclonal specifically detects GFP in Multi-Species samples. It is validated for Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GFP
Monoclonal
Unconjugated
Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
eGFP, Enhanced Green Fluorescent Protein, GFPuv, Green Fluorescent Protein, green fluorescent protein (gfp)
E.coli-derived recombinant GFPuv Ser2-Lys238 Accession # P42212
25 μg
Primary
Multi-species
IgG1
Flow Cytometry, Immunocytochemistry
454505R
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL
P42212
Mouse
Protein A or G purified from cell culture supernatant
RUO
Stains eGFP transfectants but not irrelevant transfectants in flow cytometry. Detects recombinant GFPuv in Western blots. For optimal Western blot results, we recommend R&D Systems Goat Anti-GFP Antigen Affinity-purified Polyclonal Antibody (Catalog # AF4240 ) or Novus Biologicals Mouse Anti-GFP Monoclonal Antibody (Catalog # NBP2-2211 ).
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.