missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB42401R-01M
This item is not returnable.
View return policy
Description
GFP Monoclonal specifically detects GFP in Multi-Species samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications
| GFP | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. | |
| eGFP, Enhanced Green Fluorescent Protein, GFPuv, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| E.coli-derived recombinant GFPuv Ser2-Lys238 Accession No. P42212 | |
| 1 mg | |
| Primary | |
| Multi-species | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry | |
| 454505R | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL | |
| P42212 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Stains eGFP transfectants but not irrelevant transfectants in flow cytometry. Detects recombinant GFPuv in Western blots. For optimal Western blot results, we recommend R&D Systems Goat Anti-GFP Antigen Affinity-purified Polyclonal Antibody (Catalog # AF4240 ) or Novus Biologicals Mouse Anti-GFP Monoclonal Antibody (Catalog # NBP2-2211 ). | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction