missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£165.00 - £362.00
Specifications
| Antigen | GFAP |
|---|---|
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243761
|
Novus Biologicals
NB120-16997-0.25ML |
0.25 mL |
£165.00
0.25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18229130
|
Novus Biologicals
NB120-16997 |
1 mL |
£362.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GFAP Polyclonal specifically detects GFAP in Human, Mouse, Rat, Bovine, Chicken, Feline, Guinea Pig, Primate, Sheep samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GFAP | |
| Polyclonal | |
| Purified | |
| RUO | |
| 2670 | |
| Primary | |
| Store at 4°C. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| FLJ45472, GFAP astrocytes, glial fibrillary acidic protein | |
| GFAP | |
| Affinity purified | |
| GFAP |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title